Learn More
Abnova™ Human ZRF1 Full-length ORF (AAH56682.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00027000-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the M-phase phosphoprotein (MPP) family. The gene encodes a phosphoprotein with a J domain and a Myb DNA-binding domain which localizes to both the nucleus and the cytosol. The protein is capable of forming a heterodimeric complex that associates with ribosomes, acting as a molecular chaperone for nascent polypeptide chains as they exit the ribosome. This protein was identified as a leukemia-associated antigen and expression of the gene is upregulated in leukemic blasts. Also, chromosomal aberrations involving this gene are associated with primary head and neck squamous cell tumors. This gene has a pseudogene on chromosome 6. Alternatively spliced variants which encode different protein isoforms have been described. [provided by RefSeq]
Sequence: MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQSpecifications
AAH56682.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MPHOSPH11/MPP11/ZRF1/ZUO1 | |
DNAJC2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27000 | |
ZRF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLLLPSAADGRGTAITHALTSASTLCQVEPVGRWFEAFVKRRNRNASASFQELEDKKELSEESEDEELQLEEFPMLKTLDPKDWKNQDHYAVLGLGHVRYKATQRQIKAAHKAMVLKHHPDKRKAAGEPIKEGDNDYFTCITKGSPGDSYQKLILQLIHLMKCYLIQ | |
RUO | |
DNAJC2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |