missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF713 Full-length ORF (AAI48833.1, 1 a.a. - 430 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00349075-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120314

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MEEEEMNDGSQMVRSQESLTFQDVAVDFTREEWDQLYPAQKNLYRDVMLENYRNLVALGYQLCKPEVIAQLELEEEWVIERDSLLDTHPDGENRPEIKKSTTSQNISDENQTHEMIMERLAGDSFWYSILGGLWDFDYHPEFNQENHKRYLGQVTLTHKKITQERSLECNKFAENCNLNSNLMQQRIPSIKIPLNSDTQGNSIKHNSDLIYYQGNYVRETPYEYSECGKIFNQHILLTDHIHTAEKPSECGKAFSHTSSLSQPQMLLTGEKPYKCDECGKRFSQRIHLIQHQRIHTGEKPFICNGCGKAFRQHSSFTQHLRIHTGEKPYKCNQCGKAFSRITSLTEHHRLHTGEKPYECGFCGKAFSQRTHLNQHERTHTGEKPYKCNECGKAFSQSAHLNQHRKIHTREKLCEYKCEQTVRHSPSFSST
Specifications

Specifications

AAI48833.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
74.25kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ39963
ZNF713
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
349075
ZNF713 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEEEMNDGSQMVRSQESLTFQDVAVDFTREEWDQLYPAQKNLYRDVMLENYRNLVALGYQLCKPEVIAQLELEEEWVIERDSLLDTHPDGENRPEIKKSTTSQNISDENQTHEMIMERLAGDSFWYSILGGLWDFDYHPEFNQENHKRYLGQVTLTHKKITQERSLECNKFAENCNLNSNLMQQRIPSIKIPLNSDTQGNSIKHNSDLIYYQGNYVRETPYEYSECGKIFNQHILLTDHIHTAEKPSECGKAFSHTSSLSQPQMLLTGEKPYKCDECGKRFSQRIHLIQHQRIHTGEKPFICNGCGKAFRQHSSFTQHLRIHTGEKPYKCNQCGKAFSRITSLTEHHRLHTGEKPYECGFCGKAFSQRTHLNQHERTHTGEKPYKCNECGKAFSQSAHLNQHRKIHTREKLCEYKCEQTVRHSPSFSST
RUO
ZNF713
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers