missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF447 Full-length ORF (NP_076415.2, 1 a.a. - 510 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065982-P02.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MLPLEKAFASPRSSPAPPDLPTPGSAAGVQQEEPETIPERTPADLEFSRLRFREFVYQEAAGPHQTLARLHELCRQWLMPEARSKEQMLELLVLEQFLGILPDKVRPWVVAQYPESCKKAASLVEGLADVLEEPGMLLGSPAGSSSILSDGVYERHMDPLLLPGELASPSQALGAGEIPAPSETPWLSPDPLFLEQRRVREAKTEEDGPANTEQKLKSFPEDPQHLGEWGHLDPAEENLKSYRKLLLWGYQLSQPDAASRLDTEELRLVERDPQGSSLPEGGRRQESAGCACEEAAPAGVLPELPTEAPPGDALADPPSGTTEEEEEQPGKAPDPQDPQDAESDSATGSQRQSVIQQPAPDRGTAKLGTKRPHPEDGDEQSLEGVSSSGDSAGLEAGQGPGADEPGLSRGKPYACGECGEAFAWLSHLMEHHSSHGGRKRYACQGCWKTFHFSLALAEHQKTHEKEKSYALGGARGPQPSTREAQAGARAGGPPESVEGEAPPAPPEAQRSpecifications
NP_076415.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
81.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586B1122/FLJ12895/FLJ44152/MGC2427/MGC4074/MGC8682/ZNF447 | |
ZSCAN18 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
65982 | |
ZNF447 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLPLEKAFASPRSSPAPPDLPTPGSAAGVQQEEPETIPERTPADLEFSRLRFREFVYQEAAGPHQTLARLHELCRQWLMPEARSKEQMLELLVLEQFLGILPDKVRPWVVAQYPESCKKAASLVEGLADVLEEPGMLLGSPAGSSSILSDGVYERHMDPLLLPGELASPSQALGAGEIPAPSETPWLSPDPLFLEQRRVREAKTEEDGPANTEQKLKSFPEDPQHLGEWGHLDPAEENLKSYRKLLLWGYQLSQPDAASRLDTEELRLVERDPQGSSLPEGGRRQESAGCACEEAAPAGVLPELPTEAPPGDALADPPSGTTEEEEEQPGKAPDPQDPQDAESDSATGSQRQSVIQQPAPDRGTAKLGTKRPHPEDGDEQSLEGVSSSGDSAGLEAGQGPGADEPGLSRGKPYACGECGEAFAWLSHLMEHHSSHGGRKRYACQGCWKTFHFSLALAEHQKTHEKEKSYALGGARGPQPSTREAQAGARAGGPPESVEGEAPPAPPEAQR | |
RUO | |
ZSCAN18 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |