Learn More
Abnova™ Human ZNF287 Partial ORF (NP_065704.1, 231 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057336-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq]
Sequence: PEWETKAQACTPVEDMSKLTKEETHTIKLEDSYDYDDRLERRGKGGFWKIHTDERGFSLKSVLSQEYDPTEECLSKYDIYRNNFEKHSNLIVQFDTQLDNSpecifications
NP_065704.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PEWETKAQACTPVEDMSKLTKEETHTIKLEDSYDYDDRLERRGKGGFWKIHTDERGFSLKSVLSQEYDPTEECLSKYDIYRNNFEKHSNLIVQFDTQLDN | |
RUO | |
ZNF287 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57336 | |
ZNF287 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126536/MGC141923/ZKSCAN13 | |
ZNF287 | |
Recombinant | |
wheat germ expression system |