missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF213 Partial ORF (NP_004211, 133 a.a. - 242 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007760-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127925

  • 6245.00 NOK / 25µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C2H2 zinc finger proteins, such as ZNF213, have bipartite structures in which one domain binds DNA or RNA and the other modulates target gene expression.[supplied by OMIM]

Sequence: QDVPSEEAEPEAAGRGSQATGPPPTVGARRRPSVPQEQHSHSAQPPALLKEGRPGETTDTCFVSGVHGPVALGDIPFYFSREEWGTLDPAQRDLFWDIKRENSRNTTLGF
Specifications

Specifications

NP_004211
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QDVPSEEAEPEAAGRGSQATGPPPTVGARRRPSVPQEQHSHSAQPPALLKEGRPGETTDTCFVSGVHGPVALGDIPFYFSREEWGTLDPAQRDLFWDIKRENSRNTTLGF
RUO
ZNF213
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7760
ZNF213 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CR53/ZKSCAN21
ZNF213
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers