missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF148 Partial ORF (NP_068799, 695 a.a. - 794 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007707-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Sequence: NHASATSTQDFLDQVTSQKKAEAQPVHQAYQMSSFEQPFRAPYHGSRAGIATQFSTANGQVNLRGPGTSAEFSEFPLVNVNDNRAGMTSSPDATTGQTFGSpecifications
NP_068799 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NHASATSTQDFLDQVTSQKKAEAQPVHQAYQMSSFEQPFRAPYHGSRAGIATQFSTANGQVNLRGPGTSAEFSEFPLVNVNDNRAGMTSSPDATTGQTFG | |
RUO | |
ZNF148 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7707 | |
ZNF148 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BERF-1/BFCOL1/HT-BETA/ZBP-89/ZFP148/pHZ-52 | |
ZNF148 | |
Recombinant | |
wheat germ expression system |