missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZIC1 Partial ORF (NP_003403, 2 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007545-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16127675

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. Members of this family are important during development. Aberrant expression of this gene is seen in medulloblastoma, a childhood brain tumor. This gene is closely linked to the gene encoding zinc finger protein of the cerebellum 4, a related family member on chromosome 3. This gene encodes a transcription factor that can bind and transactivate the apolipoprotein E gene. [provided by RefSeq]

Sequence: LLDAGPQYPAIGVTTFGASRHHSAGDVAERDVGLGINPFADGMGAFKLNPSSHELASAGQTAFTSQAPGYAAAAALGHHHHPGHVGSYSSAAFN
Specifications

Specifications

NP_003403
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.08kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LLDAGPQYPAIGVTTFGASRHHSAGDVAERDVGLGINPFADGMGAFKLNPSSHELASAGQTAFTSQAPGYAAAAALGHHHHPGHVGSYSSAAFN
RUO
ZIC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7545
ZIC1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ZIC/ZNF201
ZIC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers