missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZFHX4 Partial ORF (NP_078997, 2 a.a. - 93 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079776-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: ETCDSPPISRQENGQSTSKLCGTTQLDNEVPEKVAGMEPDRENSSTDDNLKTDERKSEALLGFSVENAAATQVTSAKEIPCNECATSFPSLQSpecifications
NP_078997 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.86kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETCDSPPISRQENGQSTSKLCGTTQLDNEVPEKVAGMEPDRENSSTDDNLKTDERKSEALLGFSVENAAATQVTSAKEIPCNECATSFPSLQ | |
RUO | |
ZFHX4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79776 | |
ZFHX4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ16514/FLJ20980/ZFH-4/ZFH4/ZHF4 | |
ZFHX4 | |
Recombinant | |
wheat germ expression system |