missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZC3HC1 Full-length ORF (AAH28917.1, 1 a.a. - 212 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00051530-P02.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16190103

  • 4115.00 NOK / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MTDLDASFGLTSSPIPGLEGRPERLPLVPESPRRMMTRSQDATFSPGSEQAEKSPGPIVSRTRSWDSSSPVDRPEPEAASPTTRTRPVTRSMGTGDTPGLEVPSSPLRKAKRARLCSSSSSDTSSRSFFDPTSQHRDWCPWVNITLGKESRENGGTEPDASAPAEPGWKAVLTILLAHKQSSQPAETDSMSLSEKSRKVFRIFRQWESLCSC
Specifications

Specifications

AAH28917.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
1110054L24Rik/HSPC216/NIPA
ZC3HC1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
51530
ZC3HC1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTDLDASFGLTSSPIPGLEGRPERLPLVPESPRRMMTRSQDATFSPGSEQAEKSPGPIVSRTRSWDSSSPVDRPEPEAASPTTRTRPVTRSMGTGDTPGLEVPSSPLRKAKRARLCSSSSSDTSSRSFFDPTSQHRDWCPWVNITLGKESRENGGTEPDASAPAEPGWKAVLTILLAHKQSSQPAETDSMSLSEKSRKVFRIFRQWESLCSC
RUO
ZC3HC1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers