Learn More
Abnova™ Human WNK2 Partial ORF (NP_006639, 2118 a.a. - 2217 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00065268-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a cytoplasmic serine-threonine kinase that contains cysteine in place of the lysine found at the conserved ATP-binding location in subdomain II of protein kinases. Since this protein does have kinase activity, it is possible that another lysine in the kinase subdomain I can substitute for the missing conserved lysine. [provided by RefSeq]
Sequence: KGTFTDDLHKLVDEWTSKTVGAAQLKPTLNQLKQTQKLQDMEAQAGWAAPGEARAMTAPRAGVGMPRLPPAPGPLSTTVIPGAAPTLSVPTPDPESEKPDSpecifications
NP_006639 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KGTFTDDLHKLVDEWTSKTVGAAQLKPTLNQLKQTQKLQDMEAQAGWAAPGEARAMTAPRAGVGMPRLPPAPGPLSTTVIPGAAPTLSVPTPDPESEKPD | |
RUO | |
WNK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
65268 | |
WNK2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1760/NY-CO-43/P/OKcl.13/PRKWNK2/SDCCAG43 | |
WNK2 | |
Recombinant | |
wheat germ expression system |