Learn More
Abnova™ Human WDR13 Partial ORF (NP_060353.2, 391 a.a. - 484 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064743-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is widely expressed in various tissues, and located in chromosome X. The function of this gene has not been determined. [provided by RefSeq]
Sequence: GTLQLKRSFPIEQSSHPVRSIFCPLMSFRQGACVVTGSEDMCVHFFDVERAAKAAVNKLQGHSAPVLDVSFNCDESLLASSDASGMVIVWRREQSpecifications
NP_060353.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTLQLKRSFPIEQSSHPVRSIFCPLMSFRQGACVVTGSEDMCVHFFDVERAAKAAVNKLQGHSAPVLDVSFNCDESLLASSDASGMVIVWRREQ | |
RUO | |
WDR13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
64743 | |
WDR13 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779C2057/FLJ20563/MG21 | |
WDR13 | |
Recombinant | |
wheat germ expression system |