Learn More
Abnova™ Human WBP11 Full-length ORF (NP_057396.1, 1 a.a. - 641 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00051729-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a nuclear protein, which colocalizes with mRNA splicing factors and intermediate filament-containing perinuclear networks. This protein has 95% amino acid sequence identity to the mouse Wbp11 protein. It contains two proline-rich regions that bind to the WW domain of Npw38, a nuclear protein, and thus this protein is also called Npw38-binding protein NpwBP. The Npw38-NpwBP complex may function as a component of an mRNA factory in the nucleus. [provided by RefSeq]
Sequence: MGRRSTSSTKSGKFMNPTDQARKEARKRELKKNKKQRMMVRAAVLKMKDPKQIIRDMEKLDEMEFNPVQQPQLNEKVLKDKRKKLRETFERILRLYEKENPDIYKELRKLEVEYEQKRAQLSQYFDAVKNAQHVEVESIPLPDMPHAPSNILIQDIPLPGAQPPSILKKTSAYGPPTRAVSILPLLGHGVPRLPPGRKPPGPPPGPPPPQVVQMYGRKVGFALDLPPRRRDEDMLYSPELAQRGHDDDVSSTSEDDGYPEDMDQDKHDDSTDDSDTDKSDGESDGDEFVHRDNGERDNNEEKKSGLSVRFADMPGKSRKKKKNMKELTPLQAMMLRMAGQEIPEEGREVEEFSEDDDEDDSDDSEAEKQSQKQHKEESHSDGTSTASSQQQAPPQSVPPSQIQAPPMPGPPPLGPPPAPPLRPPGPPTGLPPGPPPGAPPFLRPPGMPGLRGPLPRLLPPGPPPGRPPGPPPGPPPGLPPGPPPRGPPPRLPPPAPPGIPPPRPGMMRPPLVPPLGPAPPGLFPPAPLPNPGVLSAPPNLIQRPKADDTSAATIEKKATATISAKPQITNPKAEITRFVPTALRVRRENKGATAAPQRKSEDDSAVPLAKAAPKSGPSVPVSVQTKDDVYEAFMKEMEGLLSpecifications
NP_057396.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
96.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp779M1063/NPWBP/SIPP1 | |
WBP11 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
51729 | |
WBP11 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRRSTSSTKSGKFMNPTDQARKEARKRELKKNKKQRMMVRAAVLKMKDPKQIIRDMEKLDEMEFNPVQQPQLNEKVLKDKRKKLRETFERILRLYEKENPDIYKELRKLEVEYEQKRAQLSQYFDAVKNAQHVEVESIPLPDMPHAPSNILIQDIPLPGAQPPSILKKTSAYGPPTRAVSILPLLGHGVPRLPPGRKPPGPPPGPPPPQVVQMYGRKVGFALDLPPRRRDEDMLYSPELAQRGHDDDVSSTSEDDGYPEDMDQDKHDDSTDDSDTDKSDGESDGDEFVHRDNGERDNNEEKKSGLSVRFADMPGKSRKKKKNMKELTPLQAMMLRMAGQEIPEEGREVEEFSEDDDEDDSDDSEAEKQSQKQHKEESHSDGTSTASSQQQAPPQSVPPSQIQAPPMPGPPPLGPPPAPPLRPPGPPTGLPPGPPPGAPPFLRPPGMPGLRGPLPRLLPPGPPPGRPPGPPPGPPPGLPPGPPPRGPPPRLPPPAPPGIPPPRPGMMRPPLVPPLGPAPPGLFPPAPLPNPGVLSAPPNLIQRPKADDTSAATIEKKATATISAKPQITNPKAEITRFVPTALRVRRENKGATAAPQRKSEDDSAVPLAKAAPKSGPSVPVSVQTKDDVYEAFMKEMEGLL | |
RUO | |
WBP11 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |