Learn More
Abnova™ Human VPS53 Full-length ORF (NP_060759.2, 1 a.a. - 670 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055275-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with sequence similarity to the yeast Vps53p protein. Vps53p is involved in retrograde vesicle trafficking in late Golgi. [provided by RefSeq]
Sequence: MMEEEELEFVEELEAVLQLTPEVQLAIEQVFPSQDPLDRADFNAVEYINTLFPTEQSLANIDEVVNKIRLKIRRLDDNIRTVVRGQTNVGQDGRQVKEITRDIKQLDHAKRHLTTSITTLNHLHMLAGGVDSLEAMTRRRQYGEVANLLQGVMNVLEHFHKYMGIPQIRQLSERVKAAQTELGQQILADFEEAFPSQGTKRPGGPSNVLRDACLVANILDPRIKQEIIKKFIKQHLSEYLVLFQENQDVAWLDKIDRRYAWIKRQLVDYEEKYGRMFPREWCMAERIAVEFCHVTRAELAKIMRTRAKEIEVKLLLFAIQRTTNFEGFLAKRFSGCTLTDGTLKKLESPPPSTNPFLEDEPTPEMEELATEKGDLDQPKKPKAPDNPFHGIVSKCFEPHLYVYIESQDKNLGELIDRFVADFKAQGPPKPNTDEGGAVLPSCADLFVYYKKCMVQCSQLSTGEPMIALTTIFQKYLREYAWKILSGNLPKTTTSSGGLTISSLLKEKEGSEVAKFTLEELCLICNILSTAEYCLATTQQLEEKLKEKVDVSLIERINLTGEMDTFSTVISSSIQLLVQDLDAACDPALTAMSKMQWQNVEHVGDQSPYVTSVILHIKQNVPIIRDNLASTRKYFTQFCVKFANSFIPKFITHLFKCKPISMVGAEQVRWTSpecifications
NP_060759.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
102.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ10979/HCCS1/MGC39512/hVps53L/pp13624 | |
VPS53 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55275 | |
VPS53 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MMEEEELEFVEELEAVLQLTPEVQLAIEQVFPSQDPLDRADFNAVEYINTLFPTEQSLANIDEVVNKIRLKIRRLDDNIRTVVRGQTNVGQDGRQVKEITRDIKQLDHAKRHLTTSITTLNHLHMLAGGVDSLEAMTRRRQYGEVANLLQGVMNVLEHFHKYMGIPQIRQLSERVKAAQTELGQQILADFEEAFPSQGTKRPGGPSNVLRDACLVANILDPRIKQEIIKKFIKQHLSEYLVLFQENQDVAWLDKIDRRYAWIKRQLVDYEEKYGRMFPREWCMAERIAVEFCHVTRAELAKIMRTRAKEIEVKLLLFAIQRTTNFEGFLAKRFSGCTLTDGTLKKLESPPPSTNPFLEDEPTPEMEELATEKGDLDQPKKPKAPDNPFHGIVSKCFEPHLYVYIESQDKNLGELIDRFVADFKAQGPPKPNTDEGGAVLPSCADLFVYYKKCMVQCSQLSTGEPMIALTTIFQKYLREYAWKILSGNLPKTTTSSGGLTISSLLKEKEGSEVAKFTLEELCLICNILSTAEYCLATTQQLEEKLKEKVDVSLIERINLTGEMDTFSTVISSSIQLLVQDLDAACDPALTAMSKMQWQNVEHVGDQSPYVTSVILHIKQNVPIIRDNLASTRKYFTQFCVKFANSFIPKFITHLFKCKPISMVGAEQVRWT | |
RUO | |
VPS53 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |