Learn More
Abnova™ Human VPS35 Full-length ORF (NP_060676.2, 1 a.a. - 796 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055737-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to a group of vacuolar protein sorting (VPS) genes. The encoded protein is a component of a large multimeric complex, termed the retromer complex, involved in retrograde transport of proteins from endosomes to the trans-Golgi network. The close structural similarity between the yeast and human proteins that make up this complex suggests a similarity in function. Expression studies in yeast and mammalian cells indicate that this protein interacts directly with VPS35, which serves as the core of the retromer complex. [provided by RefSeq]
Sequence: MPTTQQSPQDEQEKLLDEAIQAVKVQSFQMKRCLDKNKLMDALKHASNMLGELRTSMLSPKSYYELYMAISDELHYLEVYLTDEFAKGRKVADLYELVQYAGNIIPRLYLLITVGVVYVKSFPQSRKDILKDLVEMCRGVQHPLRGLFLRNYLLQCTRNILPDEGEPTDEETTGDISDSMDFVLLNFAEMNKLWVRMQHQGHSRDREKRERERQELRILVGTNLVRLSQLEGVNVERYKQIVLTGILEQVVNCRDALAQEYLMECIIQVFPDEFHLQTLNPFLRACAELHQNVNVKNIIIALIDRLALFAHREDGPGIPADIKLFDIFSQQVATVIQSRQDMPSEDVVSLQVSLINLAMKCYPDRVDYVDKVLETTVEIFNKLNLEHIATSSAVSKELTRLLKIPVDTYNNILTVLKLKHFHPLFEYFDYESRKSMSCYVLSNVLDYNTEIVSQDQVDSIMNLVSTLIQDQPDQPVEDPDPEDFADEQSLVGRFIHLLRSEDPDQQYLILNTARKHFGAGGNQRIRFTLPPLVFAAYQLAFRYKENSKVDDKWEKKCQKIFSFAHQTISALIKAELAELPLRLFLQGALAAGEIGFENHETVAYEFMSQAFSLYEDEISDSKAQLAAITLIIGTFERMKCFSEENHEPLRTQCALAASKLLKKPDQGRAVSTCAHLFWSGRNTDKNGEELHGGKRVMECLKKALKIANQCMDPSLQVQLFIEILNRYIYFYEKENDAVTIQVLNQLIQKIREDLPNLESSEETEQINKHFHNTLEHLRLRRESPESEGPIYEGLILSpecifications
NP_060676.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434E1211/DKFZp434P1672/FLJ10752/FLJ13588/FLJ20388/MEM3 | |
VPS35 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55737 | |
VPS35 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPTTQQSPQDEQEKLLDEAIQAVKVQSFQMKRCLDKNKLMDALKHASNMLGELRTSMLSPKSYYELYMAISDELHYLEVYLTDEFAKGRKVADLYELVQYAGNIIPRLYLLITVGVVYVKSFPQSRKDILKDLVEMCRGVQHPLRGLFLRNYLLQCTRNILPDEGEPTDEETTGDISDSMDFVLLNFAEMNKLWVRMQHQGHSRDREKRERERQELRILVGTNLVRLSQLEGVNVERYKQIVLTGILEQVVNCRDALAQEYLMECIIQVFPDEFHLQTLNPFLRACAELHQNVNVKNIIIALIDRLALFAHREDGPGIPADIKLFDIFSQQVATVIQSRQDMPSEDVVSLQVSLINLAMKCYPDRVDYVDKVLETTVEIFNKLNLEHIATSSAVSKELTRLLKIPVDTYNNILTVLKLKHFHPLFEYFDYESRKSMSCYVLSNVLDYNTEIVSQDQVDSIMNLVSTLIQDQPDQPVEDPDPEDFADEQSLVGRFIHLLRSEDPDQQYLILNTARKHFGAGGNQRIRFTLPPLVFAAYQLAFRYKENSKVDDKWEKKCQKIFSFAHQTISALIKAELAELPLRLFLQGALAAGEIGFENHETVAYEFMSQAFSLYEDEISDSKAQLAAITLIIGTFERMKCFSEENHEPLRTQCALAASKLLKKPDQGRAVSTCAHLFWSGRNTDKNGEELHGGKRVMECLKKALKIANQCMDPSLQVQLFIEILNRYIYFYEKENDAVTIQVLNQLIQKIREDLPNLESSEETEQINKHFHNTLEHLRLRRESPESEGPIYEGLIL | |
RUO | |
VPS35 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |