missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VKORC1 Full-length ORF (NP_076869.1, 1 a.a. - 163 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079001-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193983

  • 6245.00 NOK / 25µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Vitamin K is essential for blood clotting but must be enzymatically activated. This enzymatically activated form of vitamin K is a reduced form required for the carboxylation of glutamic acid residues in some blood-clotting proteins. The product of this gene encodes the enzyme that is responsible for reducing vitamin K 2,3-epoxide to the enzymatically activated form. Fatal bleeding can be caused by vitamin K deficiency and by the vitamin K antagonist warfarin, and it is the product of this gene that is sensitive to warfarin. In humans, mutations in this gene can be associated with deficiencies in vitamin-K-dependent clotting factors and, in humans and rats, with warfarin resistance. Two pseudogenes have been identified on chromosome 1 and the X chromosome. Two alternatively spliced transcripts encoding different isoforms have been described. [provided by RefSeq]

Sequence: MGSTWGSPGWVRLALCLTGLVLSLYALHVKAARARDRDYRALCDVGTAISCSRVFSSRWGRGFGLVEHVLGQDSILNQSNSIFGCIFYTLQLLLGCLRTRWASVLMLLSSLVSLAGSVYLAWILFFVLYDFCIVCITTYAINVSLMWLSFRKVQEPQGKAKRH
Specifications

Specifications

NP_076869.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EDTP308/FLJ00289/IMAGE3455200/MGC2694/MST134/MST576/VKCFD2/VKOR
VKORC1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
79001
VKORC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGSTWGSPGWVRLALCLTGLVLSLYALHVKAARARDRDYRALCDVGTAISCSRVFSSRWGRGFGLVEHVLGQDSILNQSNSIFGCIFYTLQLLLGCLRTRWASVLMLLSSLVSLAGSVYLAWILFFVLYDFCIVCITTYAINVSLMWLSFRKVQEPQGKAKRH
RUO
VKORC1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers