missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human VEGF-121, Animal-Free Recombinant Protein, PeproTech®
Click to view available options
Quantity:
2 μg
250 μg
50 μg
Unit Size:
250µg
2µg
50µg
Description
Recombinant Human VEGF 121 is a 28.4 kDa, disulfide-linked homodimeric protein consisting of two 121 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Vascular Endothelial Growth Factor-A (VEGF-A) was originally isolated from tumor cells and is produced by a wide variety of cell types. In addition to stimulating vascular growth and vascular permeability, VEGF-A may play a role in stimulating vasodilation via nitric oxide-dependent pathways. Mouse VEGF-A has several variants, one being VEGF-120. Rat and bovine VEGF are one amino acid shorter than the human factor, and the bovine and human sequences show a homology of 95%.
Specifications
Specifications
| For Use With (Application) | Functional Assay |
| Format | Lyophilized |
| Formulation | protein with no preservative |
| Molecular Weight (g/mol) | 28.4 kDa |
| Name | Human VEGF-121, Animal-Free |
| Purification Method | HPLC, SDS-PAGE |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.1 EU/ μg |
| Gene Alias | H-VEGF-121; M-VEGF-121 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction