missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human USP31 Partial ORF (NP_065769, 1254 a.a. - 1352 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057478-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16152713

  • 6245.00 NOK / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: AGGSSVKSVCKNTGDDEAERGHQPPASQQPNANTTGKEQLVTKDPASAKHSLLSARKSKSSQLDSGVPSSPGGRQSAEKSSKKLSSSMQTSARPSQKPQ
Specifications

Specifications

NP_065769
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AGGSSVKSVCKNTGDDEAERGHQPPASQQPNANTTGKEQLVTKDPASAKHSLLSARKSKSSQLDSGVPSSPGGRQSAEKSSKKLSSSMQTSARPSQKPQ
RUO
USP31
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
57478
USP31 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA1203
USP31
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers