Learn More
Abnova™ Human USP26 Partial ORF (NP_112223, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00083844-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the ubiquitin-specific processing (UBP) family of proteases and is a deubiquitinating enzyme (DUB) with His and Cys domains. It is specifically expressed in testis tissue. Mutations in this gene have been associated with Sertoli cell-only syndrome and male infertility. [provided by RefSeq]
Sequence: MAALFLRGFVQIGNCKTGISKSKEAFIEAVERKKKDRLVLYFKSGKYSTFRLSDNIQNVVLKSYRGNQNHLHLTLQNNNGLFIEGLSSTDAEQLKIFLDRVHQNEVQPPVSpecifications
NP_112223 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAALFLRGFVQIGNCKTGISKSKEAFIEAVERKKKDRLVLYFKSGKYSTFRLSDNIQNVVLKSYRGNQNHLHLTLQNNNGLFIEGLSSTDAEQLKIFLDRVHQNEVQPPV | |
RUO | |
USP26 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
83844 | |
USP26 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC120066/MGC120067/MGC120068 | |
USP26 | |
Recombinant | |
wheat germ expression system |