Learn More
Abnova™ Human UQCRB Full-length ORF (AAH05230, 1 a.a. - 111 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007381-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein which is part of the ubiquinol-cytochrome c oxidoreductase complex which contains ten nuclear-encoded and one mitochondrial-encoded subunits. The encoded protein binds ubiquinone and participates in the transfer of electrons when ubiquinone is bound. Mutations in this gene are associated with mitochondrial complex III deficiency. A pseudogene has been described on the X chromosome. [provided by RefSeq]
Sequence: MAGKQAVSASGKWLDGIRKWYYNAAGFNKLGLMRDDTIYEDEDVKEAIRRLPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIRERKEREEWAKKSpecifications
AAH05230 | |
Liquid | |
7381 | |
UQCRB (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGKQAVSASGKWLDGIRKWYYNAAGFNKLGLMRDDTIYEDEDVKEAIRRLPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIRERKEREEWAKK | |
RUO | |
UQCRB | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.95kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ92016/FLJ97033/QCR7/QP-C/QPC/UQBC/UQBP/UQPC | |
UQCRB | |
Yes | |
wheat germ expression system |