Learn More
Abnova™ Human UBXD8 Full-length ORF (NP_055428.1, 1 a.a. - 445 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00023197-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is highly expressed in peripheral blood of patients with atopic dermatitis (AD), compared to normal individuals. It may play a role in regulating the resistance to apoptosis that is observed in T cells and eosinophils of AD patients. [provided by RefSeq]
Sequence: MAAPEERDLTQEQTEKLLQFQDLTGIESMDQCRHTLEQHNWNIEAAVQDRLNEQEGVPSVFNPPPSRPLQVNTADHRIYSYVVSRPQPRGLLGWGYYLIMLPFRFTYYTILDIFRFALRFIRPDPRSRVTDPVGDIVSFMHSFEEKYGRAHPVFYQGTYSQALNDAKRELRFLLVYLHGDDHQDSDEFCRNTLCAPEVISLINTRMLFWACSTNKPEGYRVSQALRENTYPFLAMIMLKDRRMTVVGRLEGLIQPDDLINQLTFIMDANQTYLVSERLEREERNQTQVLRQQQDEAYLASLRADQEKERKKREERERKRRKEEEVQQQKLAEERRRQNLQEEKERKLECLPPEPSPDDPESVKIIFKLPNDSRVERRFHFSQSLTVIHDFLFSLKESPEKFQIEANFPRRVLPCIPSEEWPNPPTLQEAGLSHTEVLFVQDLTDESpecifications
NP_055428.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ETEA/KIAA0887/UBXD8/UBXN3B | |
FAF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23197 | |
UBXD8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAPEERDLTQEQTEKLLQFQDLTGIESMDQCRHTLEQHNWNIEAAVQDRLNEQEGVPSVFNPPPSRPLQVNTADHRIYSYVVSRPQPRGLLGWGYYLIMLPFRFTYYTILDIFRFALRFIRPDPRSRVTDPVGDIVSFMHSFEEKYGRAHPVFYQGTYSQALNDAKRELRFLLVYLHGDDHQDSDEFCRNTLCAPEVISLINTRMLFWACSTNKPEGYRVSQALRENTYPFLAMIMLKDRRMTVVGRLEGLIQPDDLINQLTFIMDANQTYLVSERLEREERNQTQVLRQQQDEAYLASLRADQEKERKKREERERKRRKEEEVQQQKLAEERRRQNLQEEKERKLECLPPEPSPDDPESVKIIFKLPNDSRVERRFHFSQSLTVIHDFLFSLKESPEKFQIEANFPRRVLPCIPSEEWPNPPTLQEAGLSHTEVLFVQDLTDE | |
RUO | |
FAF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |