missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human UBOX5 Full-length ORF (NP_055763.1, 1 a.a. - 541 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022888-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136702

  • 3655.00 NOK / 10µg
Estimated Shipment: 26-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MVINLCLPQFRPRIHCNKISADGYEVENLISEDLTKRSHGFRTEYFIKPPVYVTVSFPFNVEICRINIDLTAGGGQNVTGLEMYTSASSSRVSWNTPQCRTLGPAEPSVPDKEAFTLVGKVLLKNQSQVVFSHRGFKARPPFGAMEATLPSPAVVAQELWNKGALSLSHVAHLRICITHVTGGGIPCIKRLEVWGQPAKTCSQEVIDSILLVTSENLPQDVALQAPALPMESDCDPGDQPESQQAPSSLQKLAEIIQDVPEEFLDPITLEIMPCPMLLPSGKVIDQSTLEKCNRSEATWGRVPSDPFTGVAFTPHSQPLPHPSLKARIDHFLLQHSIPGCHLLGRAQTALAVIPSSIVLPSQKRKIEQAEHVPDSNFGVNASCFSATSPLVLPTTSEHTAKKMKATNEPSLTHMDCSTGPLSHEQKLSQSLEIALASTLGSMPSFTARLTRGQLQHLGTRGSNTSWRPGTGSEQPGSILGPECASCKRVFSPYFKKEPVYQLPCGHLLCRPCLGEKQRSLPMTCTACQRPVASQDVLRVHF
Specifications

Specifications

NP_055763.1
Liquid
22888
UBOX5 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MVINLCLPQFRPRIHCNKISADGYEVENLISEDLTKRSHGFRTEYFIKPPVYVTVSFPFNVEICRINIDLTAGGGQNVTGLEMYTSASSSRVSWNTPQCRTLGPAEPSVPDKEAFTLVGKVLLKNQSQVVFSHRGFKARPPFGAMEATLPSPAVVAQELWNKGALSLSHVAHLRICITHVTGGGIPCIKRLEVWGQPAKTCSQEVIDSILLVTSENLPQDVALQAPALPMESDCDPGDQPESQQAPSSLQKLAEIIQDVPEEFLDPITLEIMPCPMLLPSGKVIDQSTLEKCNRSEATWGRVPSDPFTGVAFTPHSQPLPHPSLKARIDHFLLQHSIPGCHLLGRAQTALAVIPSSIVLPSQKRKIEQAEHVPDSNFGVNASCFSATSPLVLPTTSEHTAKKMKATNEPSLTHMDCSTGPLSHEQKLSQSLEIALASTLGSMPSFTARLTRGQLQHLGTRGSNTSWRPGTGSEQPGSILGPECASCKRVFSPYFKKEPVYQLPCGHLLCRPCLGEKQRSLPMTCTACQRPVASQDVLRVHF
RUO
UBOX5
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
85.4kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0860/RNF37/UBCE7IP5/UIP5
UBOX5
Yes
wheat germ expression system
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers