Learn More
Abnova™ Human UBASH3B Full-length ORF (NP_116262.2, 1 a.a. - 649 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00084959-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that contains a ubiquitin associated domain at the N-terminus, an SH3 domain, and a C-terminal domain with similarities to the catalytic motif of phosphoglycerate mutase. The encoded protein was found to inhibit endocytosis of epidermal growth factor receptor (EGFR) and platelet-derived growth factor receptor. [provided by RefSeq]
Sequence: MAQYGHPSPLGMAAREELYSKVTPRRNRQQRPGTIKHGSALDVLLSMGFPRARAQKALASTGGRSVQAACDWLFSHVGDPFLDDPLPREYVLYLRPTGPLAQKLSDFWQQSKQICGKNKAHNIFPHITLCQFFMCEDSKVDALGEALQTTVSRWKCKFSAPLPLELYTSSNFIGLFVKEDSAEVLKKFAADFAAEAASKTEVHVEPHKKQLHVTLAYHFQASHLPTLEKLAQNIDVKLGCDWVATIFSRDIRFANHETLQVIYPYTPQNDDELELVPGDFIFMSPMEQTSTSEGWIYGTSLTTGCSGLLPENYITKADECSTWIFHGSYSILNTSSSNSLTFGDGVLERRPYEDQGLGETTPLTIICQPMQPLRVNSQPGPQKRCLFVCRHGERMDVVFGKYWLSQCFDAKGRYIRTNLNMPHSLPQRSGGFRDYEKDAPITVFGCMQARLVGEALLESNTIIDHVYCSPSLRCVQTAHNILKGLQQENHLKIRVEPGLFEWTKWVAGSTLPAWIPPSELAAANLSVDTTYRPHIPISKLVVSESYDTYISRSFQVTKEIISECKSKGNNILIVAHASSLEACTCQLQGLSPQNSKDFVQMVRKIPYLGFCSCEELGETGIWQLTDPPILPLTHGPTGGFNWRETLLQESpecifications
NP_116262.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
99.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1959/MGC15437/STS-1/STS1/p70 | |
UBASH3B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
84959 | |
UBASH3B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAQYGHPSPLGMAAREELYSKVTPRRNRQQRPGTIKHGSALDVLLSMGFPRARAQKALASTGGRSVQAACDWLFSHVGDPFLDDPLPREYVLYLRPTGPLAQKLSDFWQQSKQICGKNKAHNIFPHITLCQFFMCEDSKVDALGEALQTTVSRWKCKFSAPLPLELYTSSNFIGLFVKEDSAEVLKKFAADFAAEAASKTEVHVEPHKKQLHVTLAYHFQASHLPTLEKLAQNIDVKLGCDWVATIFSRDIRFANHETLQVIYPYTPQNDDELELVPGDFIFMSPMEQTSTSEGWIYGTSLTTGCSGLLPENYITKADECSTWIFHGSYSILNTSSSNSLTFGDGVLERRPYEDQGLGETTPLTIICQPMQPLRVNSQPGPQKRCLFVCRHGERMDVVFGKYWLSQCFDAKGRYIRTNLNMPHSLPQRSGGFRDYEKDAPITVFGCMQARLVGEALLESNTIIDHVYCSPSLRCVQTAHNILKGLQQENHLKIRVEPGLFEWTKWVAGSTLPAWIPPSELAAANLSVDTTYRPHIPISKLVVSESYDTYISRSFQVTKEIISECKSKGNNILIVAHASSLEACTCQLQGLSPQNSKDFVQMVRKIPYLGFCSCEELGETGIWQLTDPPILPLTHGPTGGFNWRETLLQE | |
RUO | |
UBASH3B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |