missing translation for 'onlineSavingsMsg'
Learn More

Human u-Plasminogen Activator (uPA)/ Urokinase Catalytic Domain Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

2065.00 NOK - 20530.00 NOK

Specifications

Antigen u-Plasminogen Activator/Urokinase
Clone 204212
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Immunoprecipitation, ELISA
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18756399
GREENER_CHOICE
View Documents
R&D Systems
MAB1310-SP
25 μg
2065.00 NOK
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18779428
GREENER_CHOICE
View Documents
R&D Systems
MAB1310-100
100 μg
5005.00 NOK
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18712438
GREENER_CHOICE
View Documents
R&D Systems
MAB1310-500
500 μg
20530.00 NOK
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

u-Plasminogen Activator/Urokinase Monoclonal specifically detects u-Plasminogen Activator/Urokinase in Human samples. It is validated for Western Blot, Immunoprecipitation, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

u-Plasminogen Activator/Urokinase
LYOPH
Western Blot, Immunoprecipitation, ELISA
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ATF, EC 3.4.21, EC 3.4.21.73, plasminogen activator, urokinase, PLAU, uPA, u-PA, uPlasminogen Activator, U-plasminogen activator, urinary, Urokinase, urokinase-type plasminogen activator
PLAU
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
204212
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Mouse
Human
P00749
5328
Mouse myeloma cell line NS0-derived recombinant human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain Ser21-Leu431 Accession # P00749
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human u-Plasminogen Activator (uPA)/Urokinase Catalytic Domain in direct ELISAs and Western blots. In Western blots under reducing conditions, it reacts with the catalytic domain (B chain) only. In Western blots under non-reducing conditions, it reacts with the B chain and disulfide bond-linked B and A chains. In direct ELISAs, no cross-reactivity with recombinant human (rh) Factor X, rhHGFA, rhKLK-3, -4, -5, -6, -8, -10, -11, rhThrombin, or recombinant mouse WIF-1 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.