missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSSC1 Full-length ORF (NP_003301.1, 1 a.a. - 387 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007260-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141592

  • 6245.00 NOK / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene has been reported in PMID 9403053 as one of several tumor-suppressing subtransferable fragments located in the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. Alignment of this gene to genomic sequence data suggests that this gene resides on chromosome 2 rather than chromosome 11. [provided by RefSeq]

Sequence: MEDDAPVIYGLEFQARALTPQTAETDAIRFLVGTQSLKYDNQIHIIDFDDENNIINKNVLLHQAGEIWHISASPADRGVLTTCYNRTSDSKVLTCAAVWRMPKELESGSHESPDDSSSTAQTLELLCHLDNTAHGNMACVVWEPMGDGKKIISLADNHILLWDLQESSSQAVLASSASLEGKGQLKFTSGRWSPHHNCTQVATANDTTLRGWDTRSMSQIYCIENAHGQLVRDLDFNPNKQYYLASCGDDCKVKFWDTRNVTEPVKTLEEHSHWVWNVRYNHSHDQLVLTGSSDSRVILSNMVSISSEPFGHLVDDDDISDQEDHRSEEKSKEPLQDNVIATYEEHEDSVYAVDWSSADPWLFASLSYDGRLVINRVPRALKYHILL
Specifications

Specifications

NP_003301.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TSSC1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
7260
TSSC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEDDAPVIYGLEFQARALTPQTAETDAIRFLVGTQSLKYDNQIHIIDFDDENNIINKNVLLHQAGEIWHISASPADRGVLTTCYNRTSDSKVLTCAAVWRMPKELESGSHESPDDSSSTAQTLELLCHLDNTAHGNMACVVWEPMGDGKKIISLADNHILLWDLQESSSQAVLASSASLEGKGQLKFTSGRWSPHHNCTQVATANDTTLRGWDTRSMSQIYCIENAHGQLVRDLDFNPNKQYYLASCGDDCKVKFWDTRNVTEPVKTLEEHSHWVWNVRYNHSHDQLVLTGSSDSRVILSNMVSISSEPFGHLVDDDDISDQEDHRSEEKSKEPLQDNVIATYEEHEDSVYAVDWSSADPWLFASLSYDGRLVINRVPRALKYHILL
RUO
TSSC1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers