missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TSKS Full-length ORF (AAH58862.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00060385-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16193183

  • 4115.00 NOK / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene may play a role in testicular physiology, spermatogenesis or spermiogenesis. Expression of the encoded protein is highest in the testis and down-regulated in testicular cancer. The gene is localized to the region 19q13.3 among the related RAS viral oncogene homolog (RRAS) and interferon regulatory factor 3 (IRF3) genes, which are both involved in tumorigenesis pathways and progression. [provided by RefSeq]

Sequence: MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA
Specifications

Specifications

AAH58862.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TSKS1
TSKS
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
60385
TSKS (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASVVVKTIWQSKEIHEAGDTPTGVESCSQLVPEAPRRVTSRAKGIPKKKKAVSFHGVEPQMSHQPMHWCLNLKRSSACTNVSLLNLAAMEPTDSTGTDSTVEDLSGQLTLAGPPASPTLPWDPDDADITEILSGVNSGLVRAKDSITSLKEKTNRVNQHVQSLQSECSVLSENLERRRQEAEELEGYCIQLKENCWKVTRSVEDAEIKTNVLKQNSALLEEKLRYLQQQLQDETPRRQEAELQEPEEKQEPEEKQEPEEKQKPEAGLSWNSLGPAATSQGCPGPPGSPDKPSRPHGLVPAGWGMGPRAGEGPYVSEQELQKLFTGIEELRREVSSLTARWHQEEGAVQEALRLLGGLGGRVDGFLGQWERAQREQAQTARDLQELRGRADELCTMVERSAVSVASLRSELEGLGPLKPILEEFGRQFQNSRRGPDLSMNLDRSHQGNCARCASQGSQLSTESLQQLLDRALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDHLSNLPLEGSTGTMGGGSSAGTPPKQGGSA
RUO
TSKS
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers