Learn More
Abnova™ Human TSEN2 Full-length ORF (NP_079541.1, 1 a.a. - 465 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080746-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes one of the subunits of the tRNA splicing endonuclease. This endonuclease catalyzes the first step in RNA splicing which is the removal of introns. Mutations in this gene have been associated with pontocerebellar hypoplasia type 2. A pseudogene has been identified on chromosome 4. Multiple transcript variants encoding different isoforms have been found for this gene
Sequence: MAEAVFHAPKRKRRVYETYESPLPIPFGQDHGPLKEFKIFRAEMINNNVIVRNAEDIEQLYGKGYFGKGILSRSRPSFTISDPKLVAKWKDMKTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREPLGCLQEGSGCHPTTESFEKSVREDASPLPHVCCCKQDALILQRGLHHEDGSQHIGLLHPGDRGPDHEYVLVEEAECAMSEREAAPNEELVQRNRLICRRNPYRIFEYLQLSLEEAFFLVYALGCLSIYYEKEPLTIVKLWKAFTVVQPTFRTTYMAYHYFRSKGWVPKVGLKYGTDLLLYRKGPPFYHASYSVIIELVDDHFEGSLRRPLSWKSLAALSRVSVNVSKELMLCYLIKPSTMTDKEMESPECMKRIKVQEVILSRWVSSRERSDQDDLSpecifications
NP_079541.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
79.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC2776/MGC4440/PCH2B/SEN2/SEN2L | |
TSEN2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80746 | |
TSEN2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAEAVFHAPKRKRRVYETYESPLPIPFGQDHGPLKEFKIFRAEMINNNVIVRNAEDIEQLYGKGYFGKGILSRSRPSFTISDPKLVAKWKDMKTNMPIITSKRYQHSVEWAAELMRRQGQDESTVRRILKDYTKPLEHPPVKRNEEAQVHDKLNSGMVSNMEGTAGGERPSVVNGDSGKSGGVGDPREPLGCLQEGSGCHPTTESFEKSVREDASPLPHVCCCKQDALILQRGLHHEDGSQHIGLLHPGDRGPDHEYVLVEEAECAMSEREAAPNEELVQRNRLICRRNPYRIFEYLQLSLEEAFFLVYALGCLSIYYEKEPLTIVKLWKAFTVVQPTFRTTYMAYHYFRSKGWVPKVGLKYGTDLLLYRKGPPFYHASYSVIIELVDDHFEGSLRRPLSWKSLAALSRVSVNVSKELMLCYLIKPSTMTDKEMESPECMKRIKVQEVILSRWVSSRERSDQDDL | |
RUO | |
TSEN2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |