Learn More
Abnova™ Human TRIM6 Partial ORF (NP_001003818, 282 a.a. - 381 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00117854-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A read-through transcript transcribed from this gene and TRIM34 has been observed. [provided by RefSeq]
Sequence: DVTERSEFWTLRKPEALPTKLRSMFRAPDLKRMLRVCRELTDVQSYWVDVTLNPHTANLNLVLAKNRRQVRFVGAKVSGPSCLEKHYDCSVLGSQHFSSGSpecifications
NP_001003818 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
DVTERSEFWTLRKPEALPTKLRSMFRAPDLKRMLRVCRELTDVQSYWVDVTLNPHTANLNLVLAKNRRQVRFVGAKVSGPSCLEKHYDCSVLGSQHFSSG | |
RUO | |
TRIM6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
117854 | |
TRIM6 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RNF89 | |
TRIM6 | |
Recombinant | |
wheat germ expression system |