missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TRIM52 Partial ORF (NP_116154, 201 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084851-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140907

  • 3602.46 NOK / 10µg

Estimated Shipment: 17-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LANMVQIIRQMCPTPYRGNRSNDQGMCFKHQEALKLFCEVDKEAICVVCRESRSHKQHSVLPLEEVVQEYQEIKLETTLVGILQIEQESIHSKAYNQ
Specifications

Specifications

NP_116154
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LANMVQIIRQMCPTPYRGNRSNDQGMCFKHQEALKLFCEVDKEAICVVCRESRSHKQHSVLPLEEVVQEYQEIKLETTLVGILQIEQESIHSKAYNQ
RUO
TRIM52
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
84851
TRIM52 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC16175/RNF102
TRIM52
Yes
Videos
SDS
Documents
Certificates