Learn More
Human TRIF/TICAM1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF6216
Product Code. 18072432
  • 4970.00 NOK / 100µg
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TRIF/TICAM1
Polyclonal
Unconjugated
Q8IUC6
TICAM1
E. coli-derived recombinant human TRIF/TICAM1 Lys29-Ala204 Accession # Q8IUC6
100 μg
Primary
Detects human TRIF/TICAM1 in Western blots.
Human
IgG
Western Blot, Immunocytochemistry
LYOPH
Western Blot 0.25 ug/mL, Simple Western 25 ug/mL, Immunocytochemistry 5-15 ug/mL
MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM1, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta
Goat
Affinity Purified
RUO
148022
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.