Learn More
Human Transferrin Antibody, R&D Systems™
GREENER_CHOICE
Goat Antibody
Brand:  R&D Systems AF2914-100
 View more versions of this product
Product Code. 18730848
  • 4835.00 NOK / 100µg
Description
Transferrin specifically detects Transferrin in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transferrin
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Goat
Affinity Purified
Primary
Detects Human Transferrin in ELISAs. Detects human Transferrin and Apo-Transferrin in Western blots.
Human
IgG
Western Blot
Western Blot 0.1 ug/mL
ApoTransferrin, Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, HoloTransferrin, PRO1557, PRO2086, serotransferrin, Siderophilin, TF, transferrin
Human plasma-derived
100 μg
7018
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.