Learn More
Human Transferrin Antibody, R&D Systems™
Description
Transferrin specifically detects Transferrin in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | Transferrin |
| Applications | Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. |
| Gene Alias | ApoTransferrin, Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, HoloTransferrin, PRO1557, PRO2086, serotransferrin, Siderophilin, TF, transferrin |
| Host Species | Goat |
| Immunogen | Human plasma-derived |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |