missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TPD52L2 Partial ORF (NP_003279, 107 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007165-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136895

  • 4130.00 NOK / 10µg
Estimated Shipment: 25-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: VKTSEKLGEWNEKVTQSDLYKKTQETLSQAGQKTSAALSTVGSAISRKLGDMRNSATFKSFEDRVGTIKSKVVGDRENGSDNLPSSAGSGDKPLSDPAPF
Specifications

Specifications

NP_003279
Liquid
7165
TPD52L2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
D54/DKFZp686A1765/hD54
TPD52L2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VKTSEKLGEWNEKVTQSDLYKKTQETLSQAGQKTSAALSTVGSAISRKLGDMRNSATFKSFEDRVGTIKSKVVGDRENGSDNLPSSAGSGDKPLSDPAPF
RUO
TPD52L2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers