missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TOP1 Partial ORF (NP_003277, 692 a.a. - 765 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007150-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196835

  • 6245.00 NOK / 25µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a DNA topoisomerase, an enzyme that controls and alters the topologic states of DNA during transcription. This enzyme catalyzes the transient breaking and rejoining of a single strand of DNA which allows the strands to pass through one another, thus altering the topology of DNA. This gene is localized to chromosome 20 and has pseudogenes which reside on chromosomes 1 and 22. [provided by RefSeq]

Sequence: QRLEEQLMKLEVQATDREENKQIALGTSKLNYLDPRITVAWCKKWGVPIEKIYNKTQREKFAWAIDMADEDYEF
Specifications

Specifications

NP_003277
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.88kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QRLEEQLMKLEVQATDREENKQIALGTSKLNYLDPRITVAWCKKWGVPIEKIYNKTQREKFAWAIDMADEDYEF
RUO
TOP1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7150
TOP1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TOPI
TOP1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers