missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNIK Partial ORF (AAH55427, 1 a.a. - 110 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023043-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136882

  • 4130.00 NOK / 10µg
Estimated Shipment: 29-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Germinal center kinases (GCKs), such as TNIK, are characterized by an N-terminal kinase domain and a C-terminal GCK domain that serves a regulatory function (Fu et al., 1999 [PubMed 10521462]).[supplied by OMIM]

Sequence: MKKVTDYSSSSEESESSEEEEEDGESETHDGTVAVSDIPRLIPTGAPGSNEQYNVGMVGTHGLETSHADSFSGSISREGTLMIRETSGEKKRSGHSDSNGFAGHINLPDL
Specifications

Specifications

AAH55427
Liquid
23043
TNIK (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TNIK
Wheat Germ (in vitro)
GST
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MKKVTDYSSSSEESESSEEEEEDGESETHDGTVAVSDIPRLIPTGAPGSNEQYNVGMVGTHGLETSHADSFSGSISREGTLMIRETSGEKKRSGHSDSNGFAGHINLPDL
RUO
TNIK
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers