Learn More
Abnova™ Human TNFRSF6B Partial ORF (NP_116563, 32 a.a. - 141 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008771-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the tumor necrosis factor receptor superfamily. The encoded protein is postulated to play a regulatory role in suppressing FasL- and LIGHT-mediated cell death. It acts as a decoy receptor that competes with death receptors for ligand binding. Overexpression of this gene has been noted in gastrointestinal tract tumors, and it is located in a gene-rich cluster on chromosome 20, with other potentially tumor-related genes. Two transcript variants encoding the same isoform, but differing in the 5' UTR, have been observed for this gene. [provided by RefSeq]
Sequence: ETPTYPWRDAETGERLVCAQCPPGTFVQRPCRRDSPTTCGPCPPRHYTQFWNYLERCRYCNVLCGEREEEARACHATHNRACRCRTGFFAHAGFCLEHASCPPGAGVIAPSpecifications
NP_116563 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
ETPTYPWRDAETGERLVCAQCPPGTFVQRPCRRDSPTTCGPCPPRHYTQFWNYLERCRYCNVLCGEREEEARACHATHNRACRCRTGFFAHAGFCLEHASCPPGAGVIAP | |
RUO | |
TNFRSF6B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8771 | |
TNFRSF6B (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DCR3/DJ583P15.1.1/M68/TR6 | |
TNFRSF6B | |
Recombinant | |
wheat germ expression system |