missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TMEM30A Full-length ORF (NP_060717.1, 1 a.a. - 361 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00055754-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16141763

  • 3602.46 NOK / 10µg

Estimated Shipment: 13-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MAMNYNAKDEVDGGPPCAPGGTAKTRRPDNTAFKQQRLPAWQPILTAGTVLPIFFIIGLIFIPIGIGIFVTSNNIREIEIDYTGTEPSSPCNKCLSPDVTPCFCTINFTLEKSFEGNVFMYYGLSNFYQNHRRYVKSRDDSQLNGDSSALLNPSKECEPYRRNEDKPIAPCGAIANSMFNDTLELFLIGNDSYPIPIALKKKGIAWWTDKNVKFRNPPGGDNLEERFKGTTKPVNWLKPVYMLDSDPDNNGFINEDFIVWMRTAALPTFRKLYRLIERKSDLHPTLPAGRYSLNVTYNYPVHYFDGRKRMILSTISWMGGKNPFLGIAYIAVGSISFLLGVVLLVINHKYRNSSNTADITI
Specifications

Specifications

NP_060717.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
67.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C6orf67/CDC50A/FLJ10856
TMEM30A
Yes
wheat germ expression system
Liquid
55754
TMEM30A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAMNYNAKDEVDGGPPCAPGGTAKTRRPDNTAFKQQRLPAWQPILTAGTVLPIFFIIGLIFIPIGIGIFVTSNNIREIEIDYTGTEPSSPCNKCLSPDVTPCFCTINFTLEKSFEGNVFMYYGLSNFYQNHRRYVKSRDDSQLNGDSSALLNPSKECEPYRRNEDKPIAPCGAIANSMFNDTLELFLIGNDSYPIPIALKKKGIAWWTDKNVKFRNPPGGDNLEERFKGTTKPVNWLKPVYMLDSDPDNNGFINEDFIVWMRTAALPTFRKLYRLIERKSDLHPTLPAGRYSLNVTYNYPVHYFDGRKRMILSTISWMGGKNPFLGIAYIAVGSISFLLGVVLLVINHKYRNSSNTADITI
RUO
TMEM30A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates