missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TMEM102 (aa 103-201) Control Fragment Recombinant Protein

Product Code. 30202010
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
30202010 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 30202010

Brand: Invitrogen™ RP93484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55077 (PA5-55077. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Selectively involved in CSF2 deprivation-induced apoptosis via a mitochondria-dependent pathway.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N9M5
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 284114
Name Human TMEM102 (aa 103-201) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 4632403M07Rik; Cbap; common beta-chain associated protein; common beta-chain-associated protein; LOW QUALITY PROTEIN: transmembrane protein 102; RGD1559450; TMEM102; Tmem102-ps; transmembrane protein 102; transmembrane protein 102, pseudogene
Common Name TMEM102
Gene Symbol TMEM102
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HAPLGPYARGPHYDAGFTLLVPMFSLDGTELQLDLESCYAQVCLPEMVCGTPIREMWQDCLGPPVPGARDSIHRTESEESSKDWQSSVDQPHSYVTEHE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.