Learn More
Abnova™ Human TM2D3 Full-length ORF (AAH08873.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00080213-P01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene contains a structural module related to that of the seven transmembrane domain G protein-coupled receptor superfamily. This protein has sequence and structural similarities to the beta-amyloid binding protein (BBP), but, unlike BBP, it does not regulate a response to beta-amyloid peptide. This protein may have regulatory roles in cell death or proliferation signal cascades. Several alternatively spliced transcript variants of this gene are described but the full length nature of some variants has not been determined. Multiple polyadenylation sites have been found in this gene. [provided by RefSeq]
Sequence: MAGGVRPLRGLRALCRVLLFLSQFCILSGGESTEIPPYVMKCPSNGLCSRLPADCIDCTTNFSCTYGKPVTFDCAVKPSVTCVDQDFKSQKNFIINMTCRFCWQLPETDYECTNSTSCMTVSCPRQRYPANCTVRDHVHCLGNRTFPKMLYCNWTGGYKWSTALALSITLGGFGADRFYLGQWREGLGKLFSFGGLGIWTLIDVLLIGVGYVGPADGSLYISpecifications
AAH08873.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BLP2 | |
TM2D3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
80213 | |
TM2D3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGGVRPLRGLRALCRVLLFLSQFCILSGGESTEIPPYVMKCPSNGLCSRLPADCIDCTTNFSCTYGKPVTFDCAVKPSVTCVDQDFKSQKNFIINMTCRFCWQLPETDYECTNSTSCMTVSCPRQRYPANCTVRDHVHCLGNRTFPKMLYCNWTGGYKWSTALALSITLGGFGADRFYLGQWREGLGKLFSFGGLGIWTLIDVLLIGVGYVGPADGSLYI | |
RUO | |
TM2D3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |