Learn More
Abnova™ Human TLK2 Partial ORF (AAH44925, 1 a.a. - 126 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011011-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The Tousled-like kinases, first described in Arabidopsis, are nuclear serine/threonine kinases that are potentially involved in the regulation of chromatin assembly.[supplied by OMIM]
Sequence: MEELHSLDPRRQELLEARFTGVGVSKGPLNSESSNQSLCSVGSLSDKEVETPEKKQNDQRNRKRKAEPYETSQGKGTPRGHKISDYFEFAGGSAPGTSPGRSVPPVARSSPQHSLSNPLPRRVEQPSpecifications
AAH44925 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.27kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEELHSLDPRRQELLEARFTGVGVSKGPLNSESSNQSLCSVGSLSDKEVETPEKKQNDQRNRKRKAEPYETSQGKGTPRGHKISDYFEFAGGSAPGTSPGRSVPPVARSSPQHSLSNPLPRRVEQP | |
RUO | |
TLK2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
11011 | |
TLK2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC44450/PKU-ALPHA | |
TLK2 | |
Recombinant | |
wheat germ expression system |