Learn More
ACROBiosystems Human TIE2 Protein, Fc Tag
Brand: ACROBiosystems TI2-H5255-100ug
Additional Details : Weight : 0.00010kg
Description
Tyrosine-protein kinase acts as cell-surface receptor for ANGPT1, ANGPT2 and ANGPT4, has anti-inflammatory effects by preventing the leakage of proinflammatory plasma proteins and leukocytes from blood vessels. Required for normal angiogenesis and heart development during embryogenesis, post-natal hematopoiesis. After birth, activates or inhibits angiogenesis, depending on the context. Inhibits angiogenesis and promotes vascular stability in quiescent vessels, where endothelial cells have tight contacts.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 107.5 kDa.Specifications
107.5 kDa | |
Human TIE2, Fc Tag (TI2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Leu 748 (Accession # Q02763-1). | |
-20°C | |
TIE2 | |
Human | |
HEK293 | |
Other Recombinant Proteins |
100 μg | |
Ala 23 - Leu 748 | |
TIE2,Tie-2,TEK,VMCM, VMCM1,CD202b,Angiopoietin-1 receptor | |
Unconjugated | |
Fc Tag | |
Powder |