Learn More
ACROBiosystems Human TIE2 Protein, Fc Tag
Brand:  ACROBiosystems TI2-H5255-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17206580
  • 33576.41 NOK / 1mg
Estimated Shipment: 10-05-2024
Description
Tyrosine-protein kinase acts as cell-surface receptor for ANGPT1, ANGPT2 and ANGPT4, has anti-inflammatory effects by preventing the leakage of proinflammatory plasma proteins and leukocytes from blood vessels. Required for normal angiogenesis and heart development during embryogenesis, post-natal hematopoiesis. After birth, activates or inhibits angiogenesis, depending on the context. Inhibits angiogenesis and promotes vascular stability in quiescent vessels, where endothelial cells have tight contacts.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 107.5 kDa.
Specifications
Specifications
107.5 kDa
Human TIE2, Fc Tag (TI2-H5255) is expressed from human 293 cells (HEK293). It contains AA Ala 23 - Leu 748 (Accession # Q02763-1).
-20°C
TIE2
Human
HEK293
Other Recombinant Proteins
1 mg
Ala 23 - Leu 748
TIE2,Tie-2,TEK,VMCM, VMCM1,CD202b,Angiopoietin-1 receptor
Unconjugated
Fc Tag
Powder