Learn More
Abnova™ Human THRSP Partial ORF (NP_003242.1, 1 a.a. - 84 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007069-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is similar to the gene product of S14, a rat gene whose expression is limited to liver and adipose tissue and is controlled by nutritional and hormonal factors. This gene has been shown to be expressed in liver and adipocytes, particularly in lipomatous modules. It is also found to be expressed in lipogenic breast cancers, which suggests a role in controlling tumor lipid metabolism. [provided by RefSeq]
Sequence: MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKVSpecifications
NP_003242.1 | |
Liquid | |
7069 | |
THRSP (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC21659/S14/SPOT14 | |
THRSP | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.98kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MQVLTKRYPKNCLLTVMDRYAAEVHNMEQVVMIPSLLRDVQLSGPGGQAQAEAPDLYTYFTMLKAICVDVDHGLLPREEWQAKV | |
RUO | |
THRSP | |
Wheat Germ (in vitro) | |
GST |