missing translation for 'onlineSavingsMsg'
Learn More

Human Thrombospondin-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18799468
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
Product Code. Quantity unitSize
18799468 25 μg 25µg
18741509 100 μg 100µg
18751249 500 μg 500µg
3 options
This item is not returnable. View return policy

Product Code. 18799468

Brand: R&D Systems MAB3074SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 3 publications

Thrombospondin-1 Monoclonal specifically detects Thrombospondin-1 in Human samples. It is validated for Western Blot, Immunoprecipitation, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Thrombospondin-1
Applications Western Blot, Immunoprecipitation, ELISA
Classification Monoclonal
Clone 301221
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. CAA32889
Gene Alias THBS, THBS1, THBS-1, thrombospondin 1, Thrombospondin1, thrombospondin-1p180, TSP-1, TSP1thrombospondin-1, TSPTSP-1
Gene Symbols THBS1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-1 Asn19-Pro1170 Accession # CAA32889
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7057
Test Specificity Detects human Thrombospondin-1 in ELISAs and Western blots. In sandwich immunoassays and Western blots, no cross-reactivity with recombinant human (rh) Thrombospindin-2 or rhThrombospondin-4 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.