missing translation for 'onlineSavingsMsg'
Learn More

Human TfR (Transferrin R) Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

Brand:  R&D Systems MAB2474-500

 View more versions of this product

Product Code. 18711407

  • ONLINE EXCLUSIVE PRICE
    20305.00 NOK
    19205.55 NOK / 500µg
    Save 1099.45 NOK
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TfR (Transferrin R)
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
TFRC
Human TfR
500 μg
Primary
Detects human TfR in direct ELISAs.
Human
Supernatant
Flow Cytometry, CyTOF
29806
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7037
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.