Learn More
Abnova™ Human TFF2 Full-length ORF (AAH32820, 1 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00007032-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Members of the trefoil family are characterized by having at least one copy of the trefoil motif, a 40-amino acid domain that contains three conserved disulfides. They are stable secretory proteins expressed in gastrointestinal mucosa. Their functions are not defined, but they may protect the mucosa from insults, stabilize the mucus layer and affect healing of the epithelium. The encoded protein inhibits gastric acid secretion. This gene and two other related trefoil family member genes are found in a cluster on chromosome 21. [provided by RefSeq]
Sequence: MGRRDAQLLAALLVLGLCALAGSEKPSPCQCSRLSPHNRTNCGFPGITSDQCFDNGCCFDSSVTGVPWCFHPLPKQESDQCVMEVSDRRNCGYPGISPEECASRKCCFSNFIFEVPWCFFPKSVEDCHYSpecifications
AAH32820 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
39.93kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SML1/SP | |
TFF2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
7032 | |
TFF2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGRRDAQLLAALLVLGLCALAGSEKPSPCQCSRLSPHNRTNCGFPGITSDQCFDNGCCFDSSVTGVPWCFHPLPKQESDQCVMEVSDRRNCGYPGISPEECASRKCCFSNFIFEVPWCFFPKSVEDCHY | |
RUO | |
TFF2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |