Learn More
Abnova™ Human TCF12 Partial ORF (NP_996919, 364 a.a. - 453 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006938-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the basic helix-loop-helix (bHLH) E-protein family that recognizes the consensus binding site (E-box) CANNTG. This encoded protein is expressed in many tissues, among them skeletal muscle, thymus, B- and T-cells, and may participate in regulating lineage-specific gene expression through the formation of heterodimers with other bHLH E-proteins. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined. [provided by RefSeq]
Sequence: VGSPSPLTGTSQWPRPGGQAPSSPSYENSLHSLKNRVEQQLHEHLQDAMSFLKDVCEQSRMEDRLDRLDDAIHVLRNHAVGPSTSLPAGHSpecifications
NP_996919 | |
Liquid | |
6938 | |
TCF12 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HEB/HTF4/HsT17266/bHLHb20 | |
TCF12 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.64kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VGSPSPLTGTSQWPRPGGQAPSSPSYENSLHSLKNRVEQQLHEHLQDAMSFLKDVCEQSRMEDRLDRLDDAIHVLRNHAVGPSTSLPAGH | |
RUO | |
TCF12 | |
Wheat Germ (in vitro) | |
GST |