missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBX21 Partial ORF (AAH39739.1, 387 a.a. - 486 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00030009-Q02.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16146266

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. This gene is the human ortholog of mouse Tbx21/Tbet gene. Studies in mouse show that Tbx21 protein is a Th1 cell-specific transcription factor that controls the expression of the hallmark Th1 cytokine, interferon-gamma (IFNG). Expression of the human ortholog also correlates with IFNG expression in Th1 and natural killer cells, suggesting a role for this gene in initiating Th1 lineage development from naive Th precursor cells. [provided by RefSeq]

Sequence: LGAPRDHSYEAEFRAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPE
Specifications

Specifications

AAH39739.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue
LGAPRDHSYEAEFRAVSMKPAFLPSAPGPTMSYYRGQEVLAPGAGWPVAPQYPPKMGPASWFRPMRTLPMEPGPGGSEGRGPEDQGPPLVWTEIAPIRPE
RUO
TBX21
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
30009
TBX21 (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
T-PET/T-bet/TBET/TBLYM
TBX21
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers