missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TBX20 Full-length ORF (NP_065150.1, 1 a.a. - 297 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00057057-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16132453

  • 6245.00 NOK / 25µg
Estimated Shipment: 02-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MEFTASPKPQLSSRANAFSIAALMSSGGSKEKEATENTIKPLEQFVEKSSCAQPLGELTSLDAHGEFGGGSGSSPSSSSLCTEPLIPTTPIIPSEEMAKIACSLETKELWDKFHELGTEMIITKSGRRMFPTIRVSFSGVDPEAKYIVLMDIVPVDNKRYRYAYHRSSWLVAGKADPPLPARLYVHPDSPFTGEQLLKQMVSFEKVKLTNNELDQHGHIILNSMHKYQPRVHIIKKKDHTASLLNLKSEEFRTFIFPETVFTAVTAYQNQLITKLKIDSNPFAKGFRDSSRLTDIER
Specifications

Specifications

NP_065150.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.6kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASD4
TBX20
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
57057
TBX20 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEFTASPKPQLSSRANAFSIAALMSSGGSKEKEATENTIKPLEQFVEKSSCAQPLGELTSLDAHGEFGGGSGSSPSSSSLCTEPLIPTTPIIPSEEMAKIACSLETKELWDKFHELGTEMIITKSGRRMFPTIRVSFSGVDPEAKYIVLMDIVPVDNKRYRYAYHRSSWLVAGKADPPLPARLYVHPDSPFTGEQLLKQMVSFEKVKLTNNELDQHGHIILNSMHKYQPRVHIIKKKDHTASLLNLKSEEFRTFIFPETVFTAVTAYQNQLITKLKIDSNPFAKGFRDSSRLTDIER
RUO
TBX20
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers