missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TAF7L Partial ORF (NP_079161.2, 231 a.a. - 332 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054457-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene is similar to a mouse gene that encodes a TATA box binding protein-associated factor, and shows testis-specific expression. [provided by RefSeq]
Sequence: KKRFRKTQKKVPDVKEMEKSSFTEYIESPDVENEVKRLLRSDAEAVSTRWEVIAEDGTKEIESQGSIPGFLISSGMSSHKQGHTSSEYDMLREMFSDSRSNNSpecifications
NP_079161.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
KKRFRKTQKKVPDVKEMEKSSFTEYIESPDVENEVKRLLRSDAEAVSTRWEVIAEDGTKEIESQGSIPGFLISSGMSSHKQGHTSSEYDMLREMFSDSRSNN | |
RUO | |
TAF7L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54457 | |
TAF7L (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23157/TAF2Q/dJ738A13.1 | |
TAF7L | |
Recombinant | |
wheat germ expression system |