missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SYNJ2 Partial ORF (NP_003889, 1396 a.a. - 1495 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008871-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Synaptojanin-2 is a ubiquitously expressed inositol polyphosphate 5-phosphatase (Malecz et al., 2000 [PubMed 11084340]).[supplied by OMIM]
Sequence: GTKAMKPEAAPLLGDYQDPFWNLLHHPKLLNNTWLSKSSDPLDSGTRSPKRDPIDPVSAGASAAKAELPPDHEHKTLGHWVTISDQEKRTALQVFDPLAKSpecifications
NP_003889 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GTKAMKPEAAPLLGDYQDPFWNLLHHPKLLNNTWLSKSSDPLDSGTRSPKRDPIDPVSAGASAAKAELPPDHEHKTLGHWVTISDQEKRTALQVFDPLAK | |
RUO | |
SYNJ2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
8871 | |
SYNJ2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
INPP5H/KIAA0348/MGC44422 | |
SYNJ2 | |
Recombinant | |
wheat germ expression system |